Learn More
Lenz Laborglasintrumente Screwthread Tubes For Glassblowers
Product Code. p-7222554
Click to view available options
Thread Size:
18 GL
25 GL
45 GL
Unit Size:
Pack of 10
Product Code. 10353661
Brand: Lenz Laborglasintrumente 01300018
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Acc. to DIN 12216 and in addition to standard
  • With DIN thread (GL)
  • Material: DURAN™
Specifications
Diameter (Metric) 16 mm
Material Borosilicate Glass (3.3)
Thickness (Metric) 1.8 mm
Packaging Quantity 10/Pk.
Thread Size 18 GL