Learn More
BOLA Screw In Stopper EX
Product Code. 15688388
Click to view available options
Port Size:
15 mm
22 mm
32 mm
Unit Size:
Each
Product Code. 15688388
Brand: BOLA B17302
Screw In Stopper EX Male Thread, Hexagonal grip
  • Made of PTFE-EX.
  • For connection to BOLA Reactor Lids.
  • For closure of non-used connectors.
  • With hexagonal gripping surface in standard wrench size.
  • Earthing is made via a connection on the Reactor Lid.
Specifications
Certifications/Compliance FDA
Autoclavable Autoclavable
Port Size 15 mm
Color White