Learn More
BOLA Screw In Connector with Ground Joint EX
Product Code. 15618398
Click to view available options
Port Size:
15 mm
24 mm
Unit Size:
Each
Product Code. 15618398
Brand: BOLA B17402
Only null left
Add to basket
Add to basket
Screw In Connector with Ground Joint EX Male Thread on ground socket.
  • Made of static dissipative PTFE-EX.
  • For connection to BOLA Reactor Lids.
  • Connection with ground socket.
  • With hexagonal gripping surface in standard wrench size.
  • Earthing is made via a connection on the reactor lid.
Specifications
Certifications/Compliance FDA
Autoclavable Autoclavable
Port Size 15 mm
Color White