Learn More
BOLA Screw In Connector with Ground Joint
Product Code. 16679253
Click to view available options
Port Size:
15 mm
22 mm
24 mm
30 mm
Unit Size:
Each
Product Code. 16679253
Brand: BOLA B15402
Only null left
Add to basket
Add to basket
Screw In Connector with Ground Joint Male thread on ground socket.
  • For connection to BOLA Reactor Lids.
  • Connector with ground socket.
  • With hexagonal gripping surface in standard wrench size.
Specifications
Certifications/Compliance FDA
Autoclavable Autoclavable
Port Size 15 mm
Color White