Learn More
BOLA Screw Caps
Click to view available options
Thread Size (Metric):
GL 14
GL 18
GL 25
GL 32
Unit Size:
Each
Description
Screw cap with handy knurl, suitable for GL threads, made of PTFE. After assembly, the product is only exposed to PTFE.
Specifications
Specifications
| Color | White |
| Sterility | Non-sterile |
| Thread Size (Metric) | GL 14 |
| Neck Finish | 98 mm |
| Certifications/Compliance | FDA |
| Disposable | Reusable |
| Autoclavable | Autoclavable |