Learn More
Fisherbrand™ Screen
Product Code. 10589391
Change view
Click to view available options
For Use With (Equipment):
Fisherbrand GD-5040 Gel Dryer System
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
10589391 Fisherbrand GD-5040 Gel Dryer System Each
12358546 Fisherbrand GD-5040 Gel Dryer System Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10589391 Supplier Fisherbrand™ Supplier No. VGDSSS
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Accessory for use with vacuum gel dryers
  • Acts in conjunction with cast-aluminum base
  • Serve as porous gel support
  • Uniformly distributesthe heat and vacuum
  • Prevent gel cracking during drying
  • Pore size of stainless steel sheet: 1mm
Specifications
For Use With (Equipment) Fisherbrand GD-5040 Gel Dryer System
Type Screen