Learn More
RETSCH Scrapers
Product Code. 10094921 Shop All Retsch Products
Click to view available options
For Use With (Equipment):
For Mill GRINDOMIX GM 200
For Mill GRINDOMIX GM 300
For Mortar Grinder RM 200
Unit Size:
Each
Product Code. 10094921
Brand: RETSCH 05.723.0001
Only null left
Specifications
Product Type Scraper
For Use With (Application) For Easier Recovery of Sticky Samples
For Use With (Equipment) For Mill GRINDOMIX GM 200