Learn More
Kimberly-Clark™ Scott™ Medical Couch Covers
Product Code. 10449092
Change view
Click to view available options
Dimensions (L x W):
37 x 59 cm
Color:
White
Unit Size:
6 rolls
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Color Unit Size
10449092 37 x 59 cm White 6 rolls
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10449092 Supplier Kimberly-Clark™ Supplier No. 6004
  • ONLINE EXCLUSIVE PRICE
    298.00€
    64.77€ / 6 rolls
    Save 233.23€
    78% Off
Estimated Shipment: 22-12-2026
Protects medical couches and maintains hygiene in a healthcare environment. Scott™ Medical Couch Covers can be used with a dispensing system to help control use and reduce wastage.
  • 2-ply couch cover
  • Strong, tear resistant and durable
  • Ideal for protecting couches and maintaining high levels of hygiene in the healthcare environment
  • Perforated
  • Can be used with a Scott™ Couch Cover Wiper Dispenser to help control use and reduce wastage
TRUSTED_SUSTAINABILITY
Specifications
Product Type Clinical Rolls
Color White
Material AIRFLEX Fabric
Sterility Non-sterile
Dimensions (L x W) 37 x 59 cm
Length (English) 14.6 in.
Length (Metric) 37 cm
Ply 2-ply
Quantity 6 rolls x 135 sheets
Width (English) 23.2 in.
Width (Metric) 59 cm