Learn More
3M™ Scotch™ Fine Line Tape
Product Code. 17802686 Shop All 3M Products
Change view
Click to view available options
Quantity:
144 Rolls/Cs. (12 Rolls/Pk. 12 Pk./Cs.)
Unit Size:
144 rolls
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17802686 144 Rolls/Cs. (12 Rolls/Pk. 12 Pk./Cs.) 144 rolls
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17802686 Supplier 3M™ Supplier No. 7000048455
  • ONLINE EXCLUSIVE PRICE
    1045.00€
    948.75€ / 144 rolls
    Save 96.25€
    9% Off
Estimated Shipment: 19-01-2027
3M™ Scotch™ Fine Line Tape is a high-performance polypropylene plastic tape that has both a low profile and high adhesion for use in painting long straight lines and sweeping curves where a crisp, sharp paint line is desired.
  • Thin tape provides excellent color line separation
  • Proven excellent for painting long straight lines and smooth curves
  • Resistant to most solvents and moisture
  • Often used as a layout tape for high value multi-color and custom painted graphics.
TRUSTED_SUSTAINABILITY
Specifications
Color Green
Material Rubber
Product Type Fine Line Tape
For Use With (Application) Masking for Painting, Paint Preparation Masking
Model 218
Quantity 144 Rolls/Cs. (12 Rolls/Pk. 12 Pk./Cs.)
Thickness (English) 5 mil
Dimensions (L x W) 60 yd. x 1/8 in.