Learn More
Eppendorf™ Scientific Shaker Universal Platforms
Product Code. p-3535258
Click to view available options
Length (English) Platform:
13 in.
16 in.
18 in.
24 in.
30 in.
Width (English) Platform:
11 in.
12 in.
18 in.
36 in.
For Use With (Equipment):
Innova 2000, Excella E1/E2
Innova 2350 Shaker
Innova 43/43R, 2300, Series 4300 and 4400, E10, E25, E25R, C10, C25/C25KC Shaker
Innova I26/I-26R Shaker
Innova Model 2150 Shaker
Innova Portable Model 2050
Innova, I-Series, Excella Shaker
Unit Size:
Each
Product Code. 11692764
Brand: Eppendorf™ PTL386
Only null left
Add to basket
Add to basket
Requires
Clamps (sold separately)
For use with New Brunswick Innova™, I-Series™ and Excella™ Shakers
  • Accept a variety of accessory clamps, test tube racks and gel trays for greater flexibility
  • Hold combinations of flasks or test tube racks
  • With pre-drilled holes
Capacities listed are the most vessels of each size that will fit in a balanced pattern.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Universal Platform
Dimensions (L x W) 24 x 36 in. (610 x 914 mm)
Length (English) 24 in.
Length (English) Platform 24 in.
Length (Metric) 610 mm
Length (Metric) Platform 61 cm
Width (English) 36 in.
Width (English) Platform 36 in.
Width (Metric) 914 mm
Width (Metric) Platform 91.4 cm
For Use With (Equipment) Innova 2350 Shaker
Includes Pre-drilled holes