Learn More
Cole-Parmer™ Scientific Industries™ Vortex-Genie™ Vortex Shaker Platform Head with Cover
Shop All Cole-Parmer Products
Click to view available options
Diameter (Metric):
7.62 cm
Unit Size:
Each
Description
- Vortex Shaker Platform Head
- with Cover
- 3 inch Diameter
- For use with Scientific Industries™ Vortex-Genie™
Specifications
Specifications
| Product Type | Vortex Shaker Platform Head with Cover |
| Diameter (Metric) | 7.62 cm |
| For Use With (Equipment) | Scientific Industries™ Vortex-Genie™ |
| Includes | Cover |