Learn More
Porvair Sciences™ Science Polystyrene White Microplate
Click to view available options
No. of Wells:
384
Unit Size:
Pack of 100
Description
- Square wells maximise liquid volume per well
- White plate prevents cross talk in light emitting assays
- Black plate gives low autofluorescence background
- Volume, total μL/well: 120
- Format: 24 x 16 wells
- Dimensions [w x d x h], mm: 3.75 x 3.75 x 11.4 (well); 85.5 x 127.5 x 14.8 (plate)
Specifications
Specifications
| Format | Standard |
| Array | 16 x 24 |
| Color | White |
| Product Type | Microplate |
| Material | Polystyrene |
| No. of Wells | 384 |
| Total Volume (Metric) Well | 120 μL |
| No. per Pack | 100 |
| Quantity | 100 Each |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.