Learn More
BRAND™ Schilling Burette Bottle
Product Code. 10115243 Shop All Brand Products
Click to view available options
:
Material: LDPE
Product Code. 10115243
Brand: BRAND™ 130260
Schilling Burette Bottle equipped with screw cap and tube bushing.
  • Constructed of low density polyethylene
  • Narrow-mouth
  • With tube bushing
TRUSTED_SUSTAINABILITY
Specifications
Closure Type Screw cap
Color Amber
For Use With (Equipment) Shilling Burette
Material LDPE
Includes Screw cap and tube brushing