missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ SCH 50911
GREENER_CHOICE

Product Code. 18085662
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18085662 10 mg 10mg
18035562 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18085662 Supplier Tocris Bioscience™ Supplier No. 0984/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective and competitive GABAB antagonist; orally bioavailable

A selective, competitive and orally active GABAB antagonist. Displays an IC50 of 1.1 μM at GABAB, approximately 60 times that of CGP 35348 (Cat. No. 1245) and no binding affinity for GABAA at concentrations up to 100 μM.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 733717-87-8
Molecular Formula C8H15NO3

Specifications

Quantity 10 mg
Formula Weight 173.21
Purity >98%
Molecular Weight (g/mol) 173.212
Chemical Name or Material SCH 50911
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.