Learn More
Merck Scepter™ 3.0 Handheld Automated Cell Counter Kit
Product Code. 17181731
Change view
Click to view available options
Cell Size:
5 to 15 μm
8 to 25 μm
Product Type:
40 μm Scepter 3.0 Cell Counter and Sensors Kit
60 μm Scepter 3.0 Cell Counter and Sensors Kit
Unit Size:
1 set
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cell Size Product Type Unit Size
17181731 8 to 25 μm 60 μm Scepter 3.0 Cell Counter and Sensors Kit 1 set
17141731 5 to 15 μm 40 μm Scepter 3.0 Cell Counter and Sensors Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17181731 Supplier Merck Supplier No. PHCC360KIT
  • ONLINE EXCLUSIVE PRICE
    4735.00€
    4493.42€ / 1 sets
    Save 241.58€
    5% Off
Estimated Shipment: 24-11-2026
The Scepter™ 3.0 Cell Counter delivers rapid cell counts storing up to 999 counts which can be transferred to Excel.
  • For research use only
  • Replaces Scepter™ 2.0 and not compatible with 2.0 sensors
  • Provides fast, convenient method for counting cells and particles
  • Compatible with both the 40 and 60 μM sensors.
  • Employs the Coulter Principle in a miniaturized, handheld, format, taking less than 30 seconds
  • Prepare a dilution of the cell culture of interest and use the Scepter™ cell counter to aspirate a sample of this dilution
  •  Displays:
    • Cell concentration
    • Average cell size
    • Average cell volume
    • Histogram of size or volume distribution
TRUSTED_SUSTAINABILITY
Specifications
Product Type 60 μm Scepter 3.0 Cell Counter and Sensors Kit
Cell Size 8 to 25 μm
For Use With (Application) Cell Counting
Includes Scepter 3.0 Cell Counter, Test Bead Vial, Scepter 3.0 Charging Station and Pk. of Scepter 3.0 Sensors