Learn More
Merck Scepter™ 3.0 Cell Counter 40 μm Sensors
Product Code. 17131731
Change view
Click to view available options
Quantity:
250 Pk.
50 Pk.
500 Pk.
Unit Size:
Pack of 250
Pack of 50
Pack of 500
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17131731 500 Pk. Pack of 500
17111731 50 Pk. Pack of 50
17121731 250 Pk. Pack of 250
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17131731 Supplier Merck Supplier No. PHCC340500
  • ONLINE EXCLUSIVE PRICE
    1835.00€
    1630.00€ / Pack of 500
    Save 205.00€
    11% Off
Estimated Shipment: 27-11-2026
The Scepter™ 3.0 sensor is engineered with a micro-fabricated, cell-sensing zone enabling discrimination by cell size and volume- down to sub-micron and sub-pL resolution. Coupled with precision liquid handling channels and electronics, the Scepter™ sensor accurately provides cell population statistics.
  • For research use only
  • Provides fast, convenient method for counting cells and particles
  • Compatible with both the 40 and 60 μM sensors
  • Employs the Coulter Principle in a miniaturized, handheld, format enabling rapid cell counting, taking less than 30 seconds
  • Prepare a dilution of the cell culture of interest and use cell counter to aspirate a sample of this dilution into the sensor
  • Displays:
    • Cell concentration
    • Average cell size
    • Average cell volume
    • Histogram of size or volume distribution
TRUSTED_SUSTAINABILITY
Specifications
Product Type Scepter™ 3.0 Cell Counter Sensors
Cell Size 5 to 15 μm
Operating Cycles 50,000 to 1,500,000 particles/mL
Particle Size 4 to 18 μm
Quantity 500 Pk.
For Use With (Application) Cell Counting