Learn More
Tocris Bioscience™ SB 3CT
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
SB 3CT is a selective high affinity MMP2 inhibitor (Ki = 28 nM); also inhibits MMP9 (Ki = 400 nM). Active in vivo, neuroprotective and blood-brain barrier permeable.
Specifications
Specifications
| CAS | 292605-14-2 |
| Quantity | 10 mg |
| Solubility | Soluble to 100mM in DMSO, 20mM in ethanol |
| Molecular Formula | C15H14O3S2 |
| Formula Weight | 306.4g/mol |
| Color | White |
| Product Type | Inhibitor |