missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ SB 3CT

Selective high affinity MMP2 inhibitor
Brand: Tocris Bioscience™ 6088/10
This item is not returnable.
View return policy
Description
SB 3CT is a selective high affinity MMP2 inhibitor (Ki = 28 nM); also inhibits MMP9 (Ki = 400 nM). Active in vivo, neuroprotective and blood-brain barrier permeable.Specifications
| White | |
| 292605-14-2 | |
| 306.4g/mol | |
| Store at -20°C | |
| 10 mg |
| Inhibitor | |
| C15H14O3S2 | |
| Soluble to 100mM in DMSO, 20mM in ethanol | |
| Selective high affinity MMP2 inhibitor |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction