missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ SB 334867
Click to view available options
Quantity:
1 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
Description
SB 334867 is a selective non-peptide orexin OX1 receptor antagonist. pKb values are 7.2 and < 5 for inhibition of intracellular Ca2+ release in CHO cells expressing human OX1 and OX2 receptors respectively. Blocks orexin-A induced grooming and feeding following systemic administration in vivo.
Specifications
Specifications
| Chemical Name or Material | N-(2-Methyl-6-benzoxazolyl)-N'-1,5-naphthyridin-4-yl urea |
| CAS | 792173-99-0 |
| Assay Percent Range | >99% |
| Quantity | 10 mg |
| Solubility | Soluble to 100mM with gentle warming in DMSO |
| Molecular Formula | C17H13N5O2 |
| Formula Weight | Observed MW: 319.32 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction