missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ SB 216763

Description
Potent and selective, ATP-competitive glycogen synthase kinase-3 (GSK-3) inhibitor (IC50 = 34.3 nM for GSK-3α). Equally effective at inhibiting human GSK-3α and GSK-3β. Exhibits minimal activity against 24 other protein kinases (IC50 >10 μM). Stimulates glycogen synthesis in liver cells, and induces β-catenin-dependent gene transcription. Neuroprotective; also reduces pulmonary inflammation and fibrosis in a mouse model. Shown to maintain mouse embryonic stem cells in a pluripotent state.
Specifications
Specifications
| Quantity | 1 mg |
| Formula Weight | 371.22 |
| Purity | >98% |
| Molecular Weight (g/mol) | 371.217 |
| Chemical Name or Material | SB 216763 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?