Learn More
Thermo Scientific™ Savant™ SpeedVac™ Rotors for Medium Capacity Concentrators
Product Code. 11798589 Shop All Thermo Scientific Products
Click to view available options
Holds:
10 Centrifuge Tubes
100 (6 x 50mm) Tubes
100 (8 x 29 mm) Tubes
12 Scintillation Vials
12 Vials
24 Vials
32 Tubes
4 x 100 mL Pear-shaped Flasks
40 Microtubes
40 Tubes
6 x 50 mL Conical Tubes
60 Vials
64 Microtubes
72 Tubes
8 (18 x 100, 17 x 95, 16 x 100 mm) Tubes
Unit Size:
Each
Product Code. 11798589
Brand: Thermo Scientific™ RH2418
Select from one of these rotors to fit the type of sample volume and various sample formats for your specific application. Choose from various size of tubes, flasks, mircrowell plates.
SpeedVac Rotors for Medium Capacity Concentrators for use with Savant SpeedVac Concentrators.
Circular Rotors:
  • Easy to use
  • Multi-size tube holes accommodate different sample sizes
  • Require no additional accessories
  • Fit directly into your concentrator
TRUSTED_SUSTAINABILITY
Specifications
Description 24-place; 18 x 52mm mini scint vials
Type Rotor
For Use With (Equipment) SPD1030, SPD120, SPD130, SPD140DDA
Holds 24 Vials
Model RH24-18
Rotor System Traditional