Learn More
Sartorius Sterisart™ NF Sterility Testing System
BIOPHARMA
Product Code. 11730975 Shop All Sartorius Products
Change view
Click to view available options
Sterisart NF gamma; Female Luer Lock connection
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11730975 Sterisart NF gamma; Female Luer Lock connection
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11730975 Supplier Sartorius Supplier No. 16478GBD
  • ONLINE EXCLUSIVE PRICE
    685.00€
    609.45€ / Pack of 10
    Save 75.55€
    11% Off
 Restricted Item
Estimated Shipment: 09-12-2026
Provides completely closed system sterility testing of pharmaceuticals, open containers, closed containers, pre-filled syringes, powders, and medical devices. Sartorius Sterisart™ NF Sterility Testing System is optimal for use with isolators.
International pharmacopoeias require the complete sterility of pharmaceutical products that are injected into the blood stream or that otherwise enter the body below the skin surface. As a manufacturer of such products, you are required to supply proof of sterility of the final product batch. Sterisart™ NF is a completely closed system for the sterility testing of pharmaceutical products. It is based on the membrane filter method, however it eliminates the procedure of manipulating the filters. By this the main risk of a secondary contamination and false positive results is eliminated. A peristaltic pump transfers the sample into the filtration units, and after rinsing, the filtration units are filled with media and used for incubation of the filters without any contact to the environment.
  • reliable Sartochem™ membrane
  • high retention of microbes
  • low adsorption
  • high mechanical stability
  • pre-installed color-coded tube clamps
  • easy-to-read graduated marks
  • user-friendly (several practical adapters available)
  • product-/lot number identification
  • gas-impermeable packaging for protection against sterilants
TRUSTED_SUSTAINABILITY
Specifications
Pore Size 0.45 μm
Sterility Sterile
Filtration Area 15.7 cm
For Use With (Application) Sterility Testing
Includes Female luer lock connector
Max. Pressure 3 bar at 20°C
Sample Type Medical devices
Sterilization Method Gamma Irradiation
Temperature (English) Max. 122°F
Temperature (Metric) Max 50°C
Volume (English) 4 oz.
Volume (Metric) 120 mL
Closure Type No Septum