Description
- Syringe Filter Holder is recommended for the filtration of volumes of up to about 100 ml.
- In-Line Filter Holder has been designed for in-Line filtration with a pressure of up to 20 bar using 47mm Filter.
Specifications
Specifications
| Connectors | Hose nipples DN10 |
| Disposable | Reusable |
| Material | Stainless Steel |
| Sterility | Non-sterile |
| Filtration Area | 13 cm2 |
| Max. Pressure | 20 bar |
| Type | In-Line Stainless Steel Filter Holder |
| Diameter (Metric) Inside | 47 mm |