Learn More
Sartorius Printers for Secura™, Quintix™, Practum™ Analytical and Precision Weighing Balances
GREENER_CHOICE
Product Code. 14844741 Shop All Sartorius Products
Click to view available options
:
Standard Laboratory Printer
Product Code. 14844741
Brand: Sartorius YDP40
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
GLP and standard laboratory printers
Standard Lab Printer: YDP40
  • Transfer weight measurements to paper easily
  • Thermal printer with 203 x 203 dpi resolution
  • For Secura, Quintix and Practum laboratory balances
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Secura, Quintix, Practum Laboratory Balance
Electrical Requirements 240 V/50 Hz/10 amp single phase
Type Standard Thermal Line Printer