Learn More
Sartorius Diafiltration Cups for Vivaspin™ 20
Shop All Sartorius Products
Click to view available options
Quantity:
12/Pk.
Unit Size:
Pack of 12
Description
Use with Vivaspin 20
Use with Vivaspin 20
Specifications
Specifications
| Disposable | Reusable |
| Filter Type | Ultrafiltration |
| For Use With (Equipment) | Vivaspin 20 |
| Sample Volume | Up to 2 mL |
| Sterility | Non-sterile |
| Quantity | 12/Pk. |
| Type | Diafiltration Cup |