Learn More
Sartorius Balance Data Transfer Cables
GREENER_CHOICE
Product Code. p-4235188 Shop All Sartorius Products
Click to view available options
Connection Type:
RS-232 25 Pin to USB
Type:
RS232 to USB Cable
Length (English):
6.6 ft.
Unit Size:
Each
Product Code. 10647893
Brand: Sartorius YCC01USBM2
RS-232 Balance Data Transfer Cables in a variety of lengths.
RS-232 cables in a variety of lengths
  • For use with Sartorius balances
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) CPA Balance, Extended Balance, Master Pro LA Balance
Connection Type RS-232 25 Pin to USB
Length (English) 6.6 ft.
Length (Metric) 2 m
Type RS232 to USB Cable