Learn More
Sartorius Absorbent Pads
Product Code. p-4228235 Shop All Sartorius Products
Click to view available options
Packaging Type:
Packaged in 10 magazines
Packaged in petridish
Diameter (English) Outside:
1.85 in.
1.96 in.
Diameter (Metric) Outside:
4.7 cm
5 cm
50 mm
Unit Size:
Pack of 100
Pack of 1000
Product Code. 11780686
Brand: Sartorius 1540050N
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
presterilized in plastic magazines, which fit onto the Sartorius manual dispensing device
Sartorius 1.4mm thick absorbent pads are wetted with the appropriate liquid culture medium before a membrane filter is placed on them. They come presterilized in plastic magazines, which fit onto the Sartorius manual dispensing device.
The absorbent pads are available in two diameters:
  • 47mm has approx. 3ml absorption capacity
  • 50mm with approx. 3.5ml absorption capacity
TRUSTED_SUSTAINABILITY
Specifications
Product Type Nutrient Pad
Packaging Type Packaged in petridish
Diameter (English) Outside 1.96 in.
Diameter (Metric) Outside 5 cm