Learn More
R&D Systems™ SARS-CoV 3CL Protease (NP_828849.7) VersaClone cDNA
Specifications
Specifications
| Gene | SARS-CoV-2 3 CL Protease |
| Accession Number | NP_828849.7 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 937 bp |
| Gene ID (Entrez) | 43740578 |
| Gene Alias | 3 CL Proteinase, 3 C-like main protease, 3 CL-Mpro, COVID-19, M Proteinase, Main Protease, Mpro |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | SARS-CoV |
| Quantity | 10 μg |