missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mouse EGF, Recombinant Protein
E. coli Recombinant Protein
61.50€ - 70.50€
Specifications
Accession Number | P01133 |
---|---|
For Use With (Application) | Control, ELISA, Functional Assay, Immunohistochemistry, Western Blot |
Formulation | protein with 0.02% Tween 80, 140 mM NaCl, 2.7 mM KCl, 5% mannitol, 5% trehalose and no preservative; pH 7.4 |
Gene ID (Entrez) | 1950 |
Name | Mouse EGF |
Description
EGF (Epidermal growth factor) exerts its actions by binding to the EGF Receptor, a 170 kDa protein kinase. Activation of EGFR initiates diverse cellular pathways in response to toxic environmental stimuli, or to EGF binding to the receptor, the EGFR forms homo- or heterodimers with other family members. Each dimeric receptor complex initiates a distinct signaling pathway by recruiting different Src homology 2 (SH2) containing effector proteins. EGF is far and wide expressed in kidney, cerebrum, prostrate and salivary glands. EGF acts as a potent mitogenic factor and the phosphorylated receptor recruits adapter proteins like GRB2 that activates complex downstream signaling cascades. EGF activates at least 4 major downstream signaling cascades including the RAS-RAF-MEK-ERK, PI3 kinase-AKT, PLCgamma-PKC and STAT modules. Research studies suggest the protein may also play important role in activating the NF-kappa-B signaling cascade. Defects in the EGGF gene are the cause of hypomagnesemia type 4 and dysregulation has been associated with the growth and progression of certain cancers.Specifications
P01133 | |
protein with 0.02% Tween 80, 140 mM NaCl, 2.7 mM KCl, 5% mannitol, 5% trehalose and no preservative; pH 7.4 | |
Mouse EGF | |
-20°C | |
AI790464; beta-urogastrone; Egf; Epidermal growth factor; epidermal growth factor (beta-urogastrone); H-EGF; HOMG4; Pro-epidermal growth factor; Pro-epidermal growth factor precursor (EGF); URG; Urogastrone | |
Human | |
Recombinant | |
>90% |
Control, ELISA, Functional Assay, Immunohistochemistry, Western Blot | |
1950 | |
E. coli | |
RUO | |
EGF | |
Unconjugated | |
Lyophilized |