missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SARS-CoV-2 ORF3a (aa126-275), His Tag Recombinant Protein
E. coli Recombinant Protein
Brand: Invitrogen™ RP87667
Additional Details : Weight : 0.01000kg
Description
SARS-CoV ORF3a is a sodium or calcium ion protein with 3 transmembrane domains and is localized to the Golgi complex. ORF3a expression is relatively abundant in infected cells and is thought to contribute to viral pathogenesis. The exact function of ORF3a is unknown.Specifications
Control, ELISA, Western Blot | |
SARS-CoV-2 ORF3a (aa126-275), His Tag | |
E. coli | |
RUO | |
Virus | |
Recombinant | |
>95% |
PBS with no preservative; pH 7.2 | |
1 mg | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
2019-nCoV orf3a; Coronavirus; COVID; Covid 19; COVID19; COVID-19; COVID-19 orf3a; SARS; SARS Coronavirus; SARS CoV; SARS CoV2; SARS CoV-2; SARS-CoV orf3a; SARS-CoV-2; SARS-CoV-2 orf3a | |
Unconjugated | |
Liquid |