missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One Sapphire Polypropylene Full-Skirted 384-well PCR Microplates

Product Code. 10247563
Click to view available options
Color:
Clear
Unit Size:
Pack of 60
This item is not returnable. View return policy

Product Code. 10247563

Brand: Greiner Bio-One 785290

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

Suitable for adhesive sealers and heat sealing

Blue alphanumeric well coding
TRUSTED_SUSTAINABILITY

Specifications

Breakapart No
Format Standard
Lid Without Lid
Array 16 x 24
Autoclavable Not Autoclavable
Binding Type Non Binding
Color Clear
Material Polypropylene
No. of Wells 384
Volume (Metric) Working 25 μL
Sterility Non-sterile
Surface Treatment Non-treated
Well Shape Conical Bottom (V-well)
Dnase Free Yes
For Use With (Application) PCR
Max. RCF 4800 x g (swinging bucket rotor)
No. per Case 60
No. per Pack 15
Plate Compatibility Suitable for ABI
RNase Free Yes
Well Color Clear
Dimensions (L x W x H) 127.76 x 85.48 x 9.6 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.