Learn More
Greiner Bio-One Sapphire PCR Tubes with Attached Frosted Flat Cap
Product Code. 15975341
Click to view available options
Color:
Natural
Volume (Metric):
0.2 mL
Unit Size:
Pack of 10000
Product Code. 15975341
Brand: Greiner Bio-One 683201
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
For use in RT/Quantitative-PCR. Greiner Bio-One Sapphire PCR Tubes with Attached Frosted Flat Cap are comprehensive thermal cycler compatible.
  • Thin-walled for heat transfer during amplification
  • Free of detectable DNase, RNase, human DNA, endotoxin and PCR inhibitors
  • Packaging labelled with product symbols for warehousing
  • Used for sample handling
TRUSTED_SUSTAINABILITY
Specifications
Color Natural
Volume (Metric) 0.2 mL
Sterility Non-sterile
Material Polypropylene
Closure Type Flat Cap