Learn More
Skytec™ Sapphire Nano Foam+ Gloves
Product Code. 30274515
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
Unit Size:
10 pairs
Product Code. 30274515
Brand: Skytec™ SKG00079FD
Only null left
Add to basket
Add to basket
Outstanding level of cut protection and tactility on the thinest liner possible. With a nitrile foam coated palm, the second skin feeling has never been better to work in oily and wet conditions.
  • High cut resistance: Level F / A11
  • 25 gauge seamless liner offers unprecedented levels of fingertip sensitivity for precision tasks
  • Durable nitrile foam palm coating ensures good protection against oily and wet parts
  • Reinforced thumb crotch enhances durability
  • Touchscreen capabilities to allow usage with screen devices
  • Designed for easy movement and continuous wear
  • Breathable back of hand to reduce perspiration
  • Seamless knit prevents irritation
  • Elasticated wrist provides a secure fit
Specifications
Certifications/Compliance (Europe) CE II, UKCA, ANSI/ISEA 105-2016: Cut A7,Abrasion 4, EN 388:2016 + A1:2018 4X41F
Coating Material Foam Nitrile
Color Gray, Black
Cut Resistance Rating Level F
Hand Ambidextrous
Latex Free Yes
Material Tungsten
Product Type Cut-resistant Gloves
Size Small
EN388 Standard 4X41F
Liner Material HPPE, Tungsten, Nylon
Length (Metric) 215 mm