Learn More
Greiner Bio-One Sapphire 0.2 mL PCR Tubes with Attached Domed Cap
Product Code. 13054963
Click to view available options
Color:
Assorted
Unit Size:
Pack of 1000
Product Code. 13054963
Brand: Greiner Bio-One 671281
Suitable for real-time / quantitative PCR. Greiner Bio-One Sapphire 0.2mL PCR Tubes with Attached Domed Cap are free of detectable DNase, RNase, human DNA, endotoxin and PCR inhibitors.
  • Manufactured from medical grade polypropylene
  • Thin-walled for efficient heat transfer during amplification
  • Available in different colors
TRUSTED_SUSTAINABILITY
Specifications
Color Assorted
Volume (Metric) 0.2 mL
Material Polypropylene
Closure Type Domed Cap