Learn More
(-)-Santonin, TRC
Specifications
Specifications
| Chemical Name or Material | alpha-Santonin |
| CAS | 481-06-1 |
| Molecular Formula | C15 H18 O3 |
| Quantity | 5 g |
| Synonym | (3S,3aS,5aS,9bS)-3a,5,5a,9b-tetrahydro-3,5a,9-trimethylnaphtho[1,2-b]furan-2,8(3H,4H)-dione, (11S)-6α-hydroxy-3-oxo-eudesma-1,4-dien-12-oic acid γ-lactone, [3S-(3α,3aα,5aβ,9bβ)]-3a,5,5a,9b-tetrahydro-3,5a,9-trimethylnaphtho[1,2-b]furan-2,8(3H,4H)-dione, (-)-Santonin, (-)-α-Santonin, 1,2,3,4,4a,7-Hexahydro-1-hydroxy-α,4a,8-trimethyl-7-oxo-2-naphthaleneacetic acid γ-lactone, NSC 4900, Santonin, Semenen, alpha-Santonin, α-Santonin |
| InChI Formula | InChI=1S/C15H18O3/c1-8-10-4-6-15(3)7-5-11(16)9(2)12(15)13(10)18-14(8)17/h5,7-8,10,13H,4,6H2,1-3H3/t8-,10-,13-,15-/m0/s1 |
| SMILES | C[C@H]1[C@@H]2CC[C@@]3(C)C=CC(=O)C(=C3[C@H]2OC1=O)C |
| IUPAC Name | (3S,3aS,5aS,9bS)-3,5a,9-trimethyl-3a,4,5,9b-tetrahydro-3H-benzo[g][1]benzofuran-2,8-dione |
| Molecular Weight (g/mol) | 246.3 |
| Formula Weight | 246.1256 |