missing translation for 'onlineSavingsMsg'
Learn More

Sandospace R, TRC

Product Code. 30137433
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137433 500 mg 500mg
30135466 250 mg 250mg
30007041 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137433 Supplier Toronto Research Chemicals Supplier No. TRCS115305500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4156-21-2
Molecular Formula C9H5Cl2N4NaO3S
Molecular Weight (g/mol) 343.12
Synonym 2,4-Dichloro-6-(4-sulfophenylamino)triazine Sodium Salt, 2,4-Dichloro-6-(p-sulfoanilino)triazine Sodium Salt, 2,6-Dichloro-4-(4-sulfonanilino)-s-triazine Sodium Salt, Sandospace R, Sodium 2,6-dichloro-s-triazin-4-ylaminobenzene-4-sulfonate, Sodium 4-(2,4-dichloro-s-triazin-6-ylamino)benzenesulfonate, Sodium 4-(4,6-dichloro-s-triazin-2-ylamino)benzenesulfonate, Sodium 4-[(4,6-dichloro-1,3,5-triazin-2-yl)amino]benzenesulfonate, Sodium 4-[(4,6-dichloro-2-triazinyl)amino]benzenesulfonate, Sodium 4-[N-(4,6-dichloro-s-triazin-2-yl)amino]benzenesulfonate, Sodium N-(4,6-dichloro-s-triazin-2-yl)sulfanilate, Sodium p-(4,6-dichloro-2-s-triazinyl)aminobenzenesulfonate, p-(2,4-Dichloro-s-triazin-6-ylamino)benzenesulfonic Acid Sodium Salt, N-(4,6-Dichloro-s-triazin-2-yl)-sulfanilic Acid Sodium Salt, N-(4,6-Dichloro-s-triazin-2-yl)-sulfanilic Acid Monosodium Salt, 4-[(4,6-Dichloro-1,3,5-triazin-2-yl)amino]-benzenesulfonic Acid Monosodium Salt
IUPAC Name sodium;4-[(4,6-dichloro-1,3,5-triazin-2-yl)amino]benzenesulfonate
SMILES ClC1=NC(NC2=CC=C(S(=O)(O[Na])=O)C=C2)=NC(Cl)=N1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C9H6Cl2N4O3S.Na/c10-7-13-8(11)15-9(14-7)12-5-1-3-6(4-2-5)19(16,17)18;/h1-4H,(H,16,17,18)(H,12,13,14,15);/q;+1/p-1
Formula Weight 341.9357
Chemical Name or Material Sandospace R
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.