Learn More
OHAUS™ Sand Bath
Shop All Ohaus Products
Click to view available options
No. of Blocks:
1
2
4
6
Unit Size:
Each
Description
- Stainless steel sand baths are available for irregular samples when there is no suitable block available
- Low temperature covers help to reduce the air flow for additional temperature stability
Specifications
Specifications
| Holds | 6 Blocks |
| Material | Stainless Steel |
| Type | Sand Bath |
| No. of Blocks | 6 |
| Size | 6 Block |