Learn More
Thermo Scientific™ Sample Storage Starter Packs
Introduce automation into your workflow with Thermo Scientific™ sample storage starter packs
2502.50€ - 13550.90€
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
17886148
|
Thermo Scientific™
HHD-WP1D-2 |
Tubes and Decapper |
2502.50€
1 set |
In Stock
Log in to see stock. |
|||||
17866148
|
Thermo Scientific™
HHD-ST-WP1D-2 |
Tubes, Decapper, ST Reader |
3994.10€
1 set |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
17876148
|
Thermo Scientific™
HHD-HSX-WP1D-2 |
Tubes, Decapper, HSX Reader |
12229.80€
1 set |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
17896148
|
Thermo Scientific™
HHD-HSX-WP1D-505 |
0.5mL Tubes, Decapper, Reader |
13550.90€
1 set |
In Stock
Log in to see stock. |
|||||
17856148
|
Thermo Scientific™
HHD-HSX-WP1D-510 |
1.0mL Tubes, Decapper, Reader |
13550.90€
1 set |
In Stock
Log in to see stock. |
|||||
Description
Introduce automation into your workflow with Thermo Scientific™ sample storage starter packs. Choose from five different starter pack options: BASIC, ADVANCED, PREMIUM, PREMIUM 0.5 mL and PREMIUM 1.mL. All Starter Packs include Matrix™ ScrewTop Tri-coded Tubes with an 8-Channel Screw Cap Decapper. Advanced and Premium Starter Packs also include a barcode reader.
Engineered with the value of your samples in mind:
- Save 40% compared to buying the products individually.
- Choose from five different starter pack options.
- Eliminate the need to reformat non-2D cryotube samples.
- Secure sample tracking with 2D, 1D, and human readable codes.
- Cap/decap a full rack of tubes in less than 1 minute.
- Scan 96 tubes in as little as 1 second with advanced and premium starter packs.
- Help ensure quality with ISO-7 class cleanroom– manufactured tubes with sterility assurance.
Basic Starter Pack Includes:
- 8-Channel Screw Cap Decapper (1 each)
- 0.5 mL Matrix ScrewTop Tri-coded Tubes (1 case)
- 1.0 mL Matrix ScrewTop Tri-coded Tubes (1 case)
Advanced Starter Pack Includes:
- 8-Channel Screw Cap Decapper (1 each)
- VisionMate ST Single-Tube 2D Barcode Reader (1 each)
- 0.5 mL Matrix ScrewTop Tri-coded Tubes (1 case)
- 1.0 mL Matrix ScrewTop Tri-coded Tubes (1 case)
Premium Starter Pack Includes:
- 8-Channel Screw Cap Decapper (1 each)
- VisionMate HSX High Speed Barcode Reader (1 each)
- 0.5 mL Matrix ScrewTop Tri-coded Tubes (1 case)
- 1.0 mL Matrix ScrewTop Tri-coded Tubes (1 case)
Premium 0.5 mL Starter Pack Includes:
- 8-Channel Screw Cap Decapper (1 each)
- VisionMate HSX High Speed Barcode Reader (1 each)
- 0.5 mL Matrix ScrewTop Tri-coded Tubes (5 cases)
Premium 1.0 mL Starter Pack Includes:
- 8-Channel Screw Cap Decapper (1 each)
- VisionMate HSX High Speed Barcode Reader (1 each)
- 1.0 mL Matrix ScrewTop Tri-coded Tubes (5 cases)