Learn More
Velp Scientifica™ Sample Racks for Digestion Units
Product Code. 11715664 Shop All Velp Products
Click to view available options
For Use With (Equipment):
DK 42/26, DKl 42/26 Digesters
DK20 and DKL 20 Digesters
DK20, DKL20
DKL 20
DKL12
DKL12, 400 ml test tubes
DKL8
Unit Size:
Each
Product Code. 11715664
Brand: Velp Scientifica™ A00000180
Only null left
Add to basket
Add to basket
Facilitates nitrogen analysis and protein determination. Velp Scientifica™ Sample Racks for Digestion Units are for use with Velp Scientifica™ Kjeldahl Digestion Units.
  • Available with or without heat shields
  • Available to fit a number of tube sizes and counts
Specifications
Product Type Sample Rack
For Use With (Equipment) DK 42/26, DKl 42/26 Digesters