Learn More
Biochrom Sample Plate Replacement Kit
Product Code. 11330322
Click to view available options
For Use With (Equipment):
NanoVue Plus Spectrophotometer
Unit Size:
Each
Product Code. 11330322
Brand: Biochrom 28955301
Sample Plate Replacement Kit for use with NanoVue Plus Spectrophotometer.
  • The sample plates can be easily and quickly replaced should this ever become necessary
  • Sample plates can be clipped in and out by the user
  • Pathlength calibration must be performed following a change of plate
Specifications
Display No Display
For Use With (Equipment) NanoVue Plus Spectrophotometer
Shape Rectangular