Learn More
S Murray and Co™ SAMCO™ Stainless Steel Scissors
Click to view available options
Length (Metric):
115 mm
125 mm
Blade Style:
Straight
Tip Style:
Fine Point
Very Fine Point
Unit Size:
Each
Description
- Fully guaranteed in normal use and subject to stringent quality inspection procedures
Specifications
Specifications
| Product Type | Stainless Steel Scissors |
| Blade Style | Straight |
| Length (Metric) | 115 mm |
| Material | Stainless Steel |
| Description | Very fine points and open shanks |
| Tip Style | Very Fine Point |