missing translation for 'onlineSavingsMsg'
Learn More

Samarium(III) trifluoromethanesulfonate, 98%

Product Code. 11309116
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11309116

Brand: Thermo Scientific Alfa Aesar L20253.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 52093-28-4 | C3F9O9S3Sm | 597.55 g/mol

Used as catalyst for Baylis-Hillman reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as catalyst for Baylis-Hillman reaction.

Solubility
Insoluble in water.

Notes
Hygroscopic. Store in a cool, dry place in a tightly closed containers. Stable under recommended storage conditions. Incompatible with oxidizing agents and moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52093-28-4
Molecular Formula C3F9O9S3Sm
Molecular Weight (g/mol) 597.55
MDL Number MFCD00192431
InChI Key DDCWGUIPLGMBPO-UHFFFAOYSA-K
Synonym samarium iii trifluoromethanesulfonate, samarium triflate, samarium iii triflate, samarium trifluoromethanesulfonate, trifluoromethanesulfonic acid samarium iii salt, samarium iii trifluoromethanesulfonate 97+%, sm otf 3, 3cf3o3s.sm, samarium 3+ tritriflate
PubChem CID 10145316
IUPAC Name samarium(3+);trifluoromethanesulfonate
SMILES [Sm+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F

Specifications

Quantity 25 g
Sensitivity Hygroscopic
Solubility Information Insoluble in water.
Formula Weight 597.56
Percent Purity 98%
Chemical Name or Material Samarium(III) trifluoromethanesulfonate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.