Learn More
Abnova™ Salmon Sperm DNA, Sonicated
Product Code. 16042048
Change view
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16042048 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16042048 Supplier Abnova™ Supplier No. R0014.5mL
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Salmon Sperm DNA was shared by sonication.DNA is dissolved in deionized distilled water, with a fragment size range of 300-700 base pairs.
Specifications
Content And Storage Store at -20°C
For Use With (Application) Southern Blot,Northern Blot
Product Type DNA/RNA
Quantity 5 mL