missing translation for 'onlineSavingsMsg'
Learn More

Salicylic Acid-13C6, TRC

Product Code. 30004826
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30004826 10 mg 10mg
30012706 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30004826 Supplier Toronto Research Chemicals Supplier No. TRCS08812810MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1189678-81-6
Molecular Formula 13C6 C H6 O3
Molecular Weight (g/mol) 144.08
Synonym Salicylic acid-13C6 (phenyl-13C6), 2-Carboxyphenol-13C6, 2-Hydroxybenzene-13C6-carboxylic acid, o-Carboxyphenol-13C6, o-Hydroxybenzoic acid-13C6, 2-Hydroxy-1,2,3,4,5,6-13C6-benzoic acid
IUPAC Name 2-hydroxy(1,2,3,4,5,6-^{13}C_{6})cyclohexa-1,3,5-triene-1-carboxylic acid
SMILES O[13c]1[13cH][13cH][13cH][13cH][13c]1C(=O)O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C7H6O3/c8-6-4-2-1-3-5(6)7(9)10/h1-4,8H,(H,9,10)/i1+1,2+1,3+1,4+1,5+1,6+1
Formula Weight 144.0518
Percent Purity >95%
Chemical Name or Material Salicylic Acid-13C6
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.