Learn More
Saint-Gobain Norprene™ Chemical Tubing
Product Code. 11994488
Change view
Click to view available options
Diameter (English) Inner:
0.188 in.
Unit Size:
Pack of 15
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (English) Inner unitSize
11994488 0.188 in. Pack of 15
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11994488 Supplier Saint-Gobain Supplier No. 9571611
Only null left
Specifications
Diameter (English) Inner 0.188 in.
Material Norprene
Diameter (English) Outer 5/16 in.
Overall Length (English) 50 ft.
Thickness (English) 0.0625 in.
Maximum Pressure 29 psi