Learn More
Saint Gobain Life Sciences Chemware™ PTFE Solid Balls
Product Code. p-89673
Change view
Click to view available options
Diameter (Metric):
3 mm
6 mm
9.5 mm
Unit Size:
Pack of 100
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) unitSize
12398637 3 mm Pack of 100
10049589 9.5 mm Pack of 100
10777175 6 mm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12398637 Supplier Saint Gobain Life Sciences Supplier No. D1069326
Only null left
Add to basket
Add to basket
Chemware™ PTFE Solid Balls minimize bumping in boiling.
  • For column packing and as mixing beads
  • Inert to most liquids and vapors
  • Usable to 220°C (428°F)
TRUSTED_SUSTAINABILITY
Specifications
Type Boiling Stone
Material PTFE
Diameter (Metric) 3 mm