Learn More
Saint Gobain Life Sciences Bytac™ Surface Protector
Product Code. 15690191
Click to view available options
Dimensions (L x W):
15 ft. x 12.5 in.
15 ft. x 25 in.
Material:
FEP
FEP, Aluminum
FEP, Vinyl
Unit Size:
4.5m
4.6m
5m
Each
Pack of 5
Product Code. 15690191
Brand: Saint Gobain Life Sciences D1069329
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Put the protection of FEP fluoropolymer on any surface with Bytac™ Surface Protectors.
Put the protection of FEP fluoropolymer on any surface
  • Made of an overlay of FEP film bonded to a support backing of vinyl or aluminum (backing is coated with pressure-sensitive acrylic adhesive)
  • Protect benchtops, fume hoods, temporary tables, walls/ceilings, splash screens, walls of spray booths, and other lab surfaces
  • Help protect surfaces against absorption of toxic or radioactive chemicals
  • Corrosive chemicals, paint, oil, grease, and sticky substances wipe off easily with damp sponge
  • Cut easily with scissors
  • Install easily—simply peel off protective paper from adhesive backing and apply to surface
Order Info
Bytac protector with vinyl backing recommended for temperatures up to 93°C (200°F); with aluminum foil backing, can be used up to 182°C (360°F).
TRUSTED_SUSTAINABILITY
Specifications
Product Type Bench Protector
Material FEP, Aluminum
Length (English) 15 ft.
Length (Metric) 4.6 m
Width (English) 25 in.
Width (Metric) 640 mm
Dimensions (L x W) 15 ft. x 25 in.
For Use With (Application) Protects benchtops, fume hoods, temporary tables, walls/ceilings, splash screens, walls of spray booths, and other lab surfaces
Quantity 1 Ea.