missing translation for 'onlineSavingsMsg'
Learn More

Saikosaponin C, TRC

Product Code. 30428131
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30428131 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30428131 Supplier Toronto Research Chemicals Supplier No. TRCS083910100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Saikosaponin C
CAS 20736-08-7
Molecular Formula C48 H78 O17
Quantity 100 mg
Synonym β-D-Glucopyranoside','(3β,16β)-13,28-epoxy-16-hydroxyolean-11-en-3-yl O-6-deoxy-α-L-mannopyranosyl-(1→4)-O-[β-D-glucopyranosyl-(1→6)]-, Saikosaponin C (8CI), Oleanane','β-D-glucopyranoside deriv., (3β,16β)-13,28-Epoxy-16-hydroxyolean-11-en-3-yl O-6-deoxy-α-L-mannopyranosyl-(1→4)-O-[β-D-glucopyranosyl-(1→6)]-β-D-glucopyranoside
InChI Formula InChI=1S/C48H78O17/c1-22-30(51)32(53)36(57)40(61-22)65-38-24(20-59-39-35(56)33(54)31(52)23(19-49)62-39)63-41(37(58)34(38)55)64-29-11-12-44(6)25(43(29,4)5)9-13-45(7)26(44)10-14-48-27-17-42(2,3)15-16-47(27,21-60-48)28(50)18-46(45,48)8/h10,14,22-41,49-58H,9,11-13,15-21H2,1-8H3/t22-,23+,24+,25-,26+,27+,28-,29-,30-,31+,32+,33-,34+,35+,36+,37+,38+,39+,40-,41-,44-,45+,46-,47+,48-/m0/s1
SMILES C[C@@H]1O[C@@H](O[C@H]2[C@H](O)[C@@H](O)[C@H](O[C@H]3CC[C@@]4(C)[C@@H](CC[C@]5(C)[C@@H]4C=C[C@]67OC[C@@]8(CCC(C)(C)C[C@@H]68)[C@@H](O)C[C@@]57C)C3(C)C)O[C@@H]2CO[C@@H]9O[C@H](CO)[C@@H](O)[C@H](O)[C@H]9O)[C@H](O)[C@H](O)[C@H]1O
Molecular Weight (g/mol) 927.12
Formula Weight 926.5239
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.