Learn More
R&D Systems™ SAH-SOS1A
Description
SAH-SOS1A is a KRas/son of sevenless 1 (SOS1) interaction inhibitor. Binds within nucleotide binding pocket of KRas (Kd values are 106 - 176 nM for wild type KRas and various KRas mutants). Inhibits nucleotide binding to KRas in a concentration dependent manner. Displays cytotoxicity in KRas-driven cancer cell lines and inhibits downstream ERK-MAPK signaling. Cell permeable.
Sold under license from Dana-Farber Cancer Institute
Specifications
Specifications
| Product Line | Tocris |
| CAS | 1652561-87-9 |
| Content And Storage | Store at -20°C |
| Description | KRas/SOS1 interaction inhibitor |
| Formulation | C100H159N27O28 |
| Molecular Weight (g/mol) | 2187.53 |
| Quantity | 500 μg |
| Sequence | RRFFGIXLTNXLKTEEGN (Modifications: Arg-1 = N-terminal Ac, X = (S)-2-(4-pentenyl)alanine, X-7 and X-11 stapled together with a double bond) |