Learn More
Tocris Bioscience™ SAG 21k
Description
SAG 21k is a potent Hedgehog signaling activator (EC50 = 0.4 nM). Orally bioavailable and brain penetrant.
Specifications
Specifications
| Chemical Name or Material | 3-Chloro-4,7-difluoro-N-[[2-methoxy-5-(4-pyridinyl)phenyl]methyl]-N-trans-4-(methylamino)cyclohexyl]benzo[b]thiophene-2-carboxamide |
| CAS | 946002-48-8 |
| Quantity | 10 mg |
| Solubility | Soluble to 100mM in DMSO and to 20mM in ethanol |
| Molecular Formula | C29H28ClF2N3O2S |
| Formula Weight | 556.07 |
| Physical Form | Solid |
| Color | White |
| Product Type | Bioactive Small Molecule |
| Purity | >98% |
For Research Use Only.